Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle
108-90-7
C6H5Cl
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl
108-90-7
C6H5Cl
112.556
MVPPADPHJFYWMZ-UHFFFAOYSA-N
monochlorobenzene,benzene chloride,phenyl chloride,benzene, chloro,chlorbenzene,chlorobenzol,monochlorbenzol,chlorbenzol,chlorobenzen,clorobenzene
7964
CHEBI:28097
chlorobenzene
C1=CC=C(C=C1)Cl

High quality chemicals for laboratory and specialized industrial use

Formula Weight 112.56
Grade BAKER ANALYZED™ ACS Reagent
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.11kg
Flash Point 28.00°C
Quantity 4L
Chemical Name or Material Chlorobenzene