Alfa Aesar™ Chlorobenzene-d5, 99% (Isotopic)

Manufacturer: Alfa Aesar™ 4471203

Quantity 1g

 View more versions of this product

Catalog No. AA4471203

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3114-55-4
    Molecular Formula C6D5Cl
    Formula Weight 117.6
    MDL Number MFCD00000531
    UN Number UN1134
    Beilstein 1866335
    Chemical Name or Material Chlorobenzene-d5, Isotopic

    Specifications

    Formula Weight 117.6
    Refractive Index 1.522
    MDL Number MFCD00000531
    Beilstein 1866335
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H226-H332
    Flammable liquid and vapor.
    Harmful if inhaled.
    Quantity 1g
    Flash Point 24°C (75°F)
    Odor Aromatic
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: CHLOROBENZENE
    Density 1.157g/mL
    UN Number UN1134
    Health Hazard 3 GHS P Statement
    P262-P273
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.
    Molecular Formula C6D5Cl
    CAS 3114-55-4
    Boiling Point 130°C
    Physical Form Liquid
    Chemical Name or Material Chlorobenzene-d5, Isotopic
    Assay 99%

    GHS H Statement
    H226-H332
    Flammable liquid and vapor.
    Harmful if inhaled.

    GHS P Statement
    P262-P273
    Do not get in eyes, on skin, or on clothing.
    Avoid release to the environment.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit