Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorobenzene, Exceeds A.C.S. Specifications, HPLC Grade, 99.5%, Spectrum™

Click to view available options
Quantity:
1 L
Packaging:
Amber Glass Bottle
108-90-7
C6H5Cl
112.56
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
ClC1=CC=CC=C1
108-90-7
C6H5Cl
112.56
MVPPADPHJFYWMZ-UHFFFAOYSA-N
chlorobenzene
ClC1=CC=CC=C1

HP492, 108-90-7, C6H5Cl

Spectrum™ Chlorobenzene, Exceeds A.C.S. Specifications, HPLC Grade is an aromatic organic compound that is colorless, and flammable. It is commonly used and solvent. All HPLC and Ultrapure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
CAS Max % 1
UN Number UN1134
Percent Purity 99.5%
Packaging Amber Glass Bottle
Titratable Acid 4 μeq/g
Quantity 1 L