Learn More
Chlorobenzene, Exceeds A.C.S. Specifications, HPLC Grade, 99.5%, Spectrum™
Click to view available options
Packaging:
Amber Glass Bottle
Quantity:
1 L
HP492, 108-90-7, C6H5Cl
Spectrum™ Chlorobenzene, Exceeds A.C.S. Specifications, HPLC Grade is an aromatic organic compound that is colorless, and flammable. It is commonly used and solvent. All HPLC and Ultrapure solvents offered by Spectrum are glass distilled, submicron filtered, and undergo rigorous specification testing to insure accurate, repeatable results.
Packaging Amber Glass Bottle
Residue after Evaporation 2 ppm
Water 0.0003