Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorobis(2-methoxyphenyl)phosphine, 98+%

Click to view available options
Quantity:
1g
5g
263369-88-6
C18H17ClO2P-
331.756
MFCD08064037
WVGQIZOJKMMSSK-UHFFFAOYSA-N
bis 2-methoxyphenyl chlorphosphine, acmc-20alnz, phosphinous chloride,p,p-bis 2-methoxyphenyl
57371085
COC1=CC=CC=C1C2=C(P[Cl-]C=C2)C3=CC=CC=C3OC
263369-88-6
C18H17ClO2P-
331.756
MFCD08064037
WVGQIZOJKMMSSK-UHFFFAOYSA-N
bis 2-methoxyphenyl chlorphosphine, acmc-20alnz, phosphinous chloride,p,p-bis 2-methoxyphenyl
57371085
COC1=CC=CC=C1C2=C(P[Cl-]C=C2)C3=CC=CC=C3OC
Applications
Chlorobis(2-methoxyphenyl)phosphine is used as a reactant for cycloadditions reactions, asymmetric hydrogenation and allylic substitution, asymmetric hydrogenation of imines and olefins. It is also used as a reactant for synthesis of ferrocene-based chiral diphosphine, catalysts for C-C cross coupling reactions, palladium complexes.

Solubility
Reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.
Sensitivity Air and moisture sensitive
Melting Point 111°C to 114°C
Quantity 1g
UN Number UN3261
Solubility Information Reacts with water.
Formula Weight 280.69
Percent Purity ≥98%
Chemical Name or Material Chlorobis(2-methoxyphenyl)phosphine

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 000-000-0

TSCA : No

Recommended Storage : Ambient temperatures; Store under Nitrogen