Alfa Aesar™ Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 98+%, may contain suspended dimethylamine hydrochloride crystals

Manufacturer: Alfa Aesar™ H2588306

Quantity 5g

 View more versions of this product

Catalog No. AAH2588306

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 142421-57-6
    Molecular Formula C16H6ClF12P
    Formula Weight 492.63
    MDL Number MFCD01630852
    UN Number UN3265
    Synonym Bis[3,5-bis(trifluoromethyl)phenyl]chlorophosphine
    Chemical Name or Material Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine

    Specifications

    Formula Weight 492.63
    MDL Number MFCD01630852
    UN Number UN3265
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Molecular Formula C16H6ClF12P
    CAS 142421-57-6
    Synonym Bis[3,5-bis(trifluoromethyl)phenyl]chlorophosphine
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Quantity 5g
    Chemical Name or Material Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine
    Assay ≥98%, may contain suspended dimethylamine hydrochloride crystals

    Description

    Applications
    Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.

    Solubility
    Reacts with water.

    Notes
    Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit