Learn More
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine, 97%, maycontain suspended amine hydrochloride crystals, Thermo Scientific Chemicals
Description
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine is used as a reactant for the synthesis of chiral phosphine-aminophosphine ligands for rhodium-catalyzed asymmetric hydrogenation, ligands for palladium-catalyzed stereoselective allylation reactions, enantioselective hydrogenations, Josiphos analog as catalyst for asymmetric hydrogenation, ligands used in asymmetric hydrovinylation reactions.
Solubility
Reacts with water.
Notes
Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.
Specifications
Quantity | 1 g |
UN Number | UN3265 |
Sensitivity | Air and moisture sensitive; May form a precipitate |
Solubility Information | Reacts with water. |
Formula Weight | 492.63 |
Percent Purity | ≥98% |
Assay | may contain suspended dimethylamine hydrochloride crystals |
Chemical Name or Material | Chlorobis[3,5-bis(trifluoromethyl)phenyl]phosphine |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.