Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorobis(4-chlorophenyl)phosphine, 97%

Manufacturer:  Alfa Aesar™ H2588606

Quantity 5g

 View more versions of this product

Catalog No. AAH2588606


  Check Availability
Add to cart

Specifications

Description

Applications
Chlorobis(4-chlorophenyl)phosphine are now employed in tuning the properties of catalyst. Also used as primary and secondary intermediates.

Solubility
Reacts with water.

Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Description & Specifications

Chemical Identifiers

13685-26-2
289.53
UN3261
Chlorobis(4-chlorophenyl)phosphine
C12H8Cl3P
MFCD09909564
Bis(4-chlorophenyl)chlorophosphine

Specifications

184° to 186°C (2.5mmHg)
5g
C12H8Cl3P
MFCD09909564
Bis(4-chlorophenyl)chlorophosphine
97%
52° to 53°C
13685-26-2
289.53
UN3261
Chlorobis(4-chlorophenyl)phosphine

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit