Alfa Aesar™ Chlorobis(4-methoxyphenyl)phosphine, 98+%

Manufacturer: Alfa Aesar™ H2588706

Quantity 5g

 View more versions of this product

Catalog No. AAH2588706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 13685-30-8
    Molecular Formula C14H14ClO2P
    Formula Weight 280.69
    MDL Number MFCD04972301
    UN Number UN3261
    Synonym Bis(4-methoxyphenyl)chlorophosphine
    Chemical Name or Material Chlorobis(4-methoxyphenyl)phosphine

    Specifications

    Formula Weight 280.69
    Odor Irritating
    Quantity 5g
    CAS 13685-30-8
    UN Number UN3261
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Molecular Formula C14H14ClO2P
    Synonym Bis(4-methoxyphenyl)chlorophosphine
    Boiling Point 140°C (1.5mmHg)
    Melting Point 61° to 64°C
    MDL Number MFCD04972301
    Health Hazard 1 Danger
    Chemical Name or Material Chlorobis(4-methoxyphenyl)phosphine
    Assay ≥98%

    Description

    Applications
    Chlorobis(4-methoxyphenyl)phosphine is used an pharmaceutical intermediate.

    Solubility
    Reacts with water.

    Notes
    Air & moisture sensitive. Store away from oxidizing agents, bases, air, water/ moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P280-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit