Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorobis[4-(trifluoromethyl)phenyl]phosphine, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
Description
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Chlorobis[4-(trifluoromethyl)phenyl]phosphine is a reactant in the synthesis of green phosphorescent OLEDs(based on a novel iridium complex).
Solubility
Reacts with water.
Notes
Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.
Specifications
Boiling Point | 110°C (0.4 mmHg) |
Quantity | 1 g |
UN Number | UN3265 |
Sensitivity | Air and moisture sensitive |
Solubility Information | Reacts with water. |
Formula Weight | 356.64 |
Percent Purity | 97% |
Chemical Name or Material | Chlorobis[4-(trifluoromethyl)phenyl]phosphine |
Safety and Handling
Hazard Category | H314-H335 |
Hazard Statement | GHS H Statement H314-H318 Causes severe skin burns and eye damage. Causes serious eye damage. |
Precautionary Statement | P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c |
DOTInformation | Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S. |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures; Store under Nitrogen |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction