Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Chlorobutanol, Hydrous, NF, 98-100.5%, Spectrum™ Chemical
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
Description
Spectrum™ Chemical Chlorobutanol, Hydrous, NF is a very effective preservative in many pharmaceutical products for eyes, ears and nose, dental preparations. The hemihydrate form is useful in formulations where the absence of water is not critical. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
CAS Max % | 100% |
Packaging | Amber Glass Bottle |
Quantity | 500 g |
Percent Purity | 98-100.5% |
Grade | NF |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction