Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorocarbonylsulfenyl chloride, 95%, Thermo Scientific Chemicals

Catalog Number p-3777859
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2757-23-5
CCl2OS
130.97
MFCD00000703
MNOALXGAYUJNKX-UHFFFAOYSA-N
chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
75990
ClSC(Cl)=O
2757-23-5
CCl2OS
130.97
MFCD00000703
MNOALXGAYUJNKX-UHFFFAOYSA-N
chlorocarbonylsulfenyl chloride, chlorothio formyl chloride, chloroformylsulfenyl chloride, chloro chlorosulfanyl methanone, chlorothioformyl chloride, chlorocarbonylsulphenyl chloride, chlorocarbonyl sulfenyl chloride, chlorocarbonyl sulphenyl chloride, carbonochloridothioic acid, anhydrosulfide with thiohypochlorous acid, methane, chloro chlorothio oxo
75990
ClSC(Cl)=O

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Color Yellow
Density 1.5800g/mL
Boiling Point 98°C to 100°C
Flash Point 62°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (GC)
Packaging Glass bottle
Linear Formula ClCOSCl
Quantity 5 g
Specific Gravity 1.58
Solubility Information Solubility in water: reacts
IUPAC Name chloro(chlorosulfanyl)methanone
Formula Weight 130.98
Percent Purity 95%
Physical Form Liquid, Fuming In Moist Air
Chemical Name or Material Chlorocarbonylsulfenyl chloride
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre
EINECSNumber 220-415-2

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit