Alfa Aesar™ Chlorodi(2-furyl)phosphine, 98+%

Manufacturer: Alfa Aesar™ H2589203

Quantity 1g

 View more versions of this product

Catalog No. AAH2589203

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Chemical Identifiers

    CAS 181257-35-2
    Molecular Formula C8H6ClO2P
    Formula Weight 200.56
    MDL Number MFCD01630851
    UN Number UN3129
    Synonym Di(2-furyl)chlorophosphine
    Chemical Name or Material Chlorodi(2-furyl)phosphine

    Specifications

    Formula Weight 200.56
    Boiling Point 102° to 104°C (14mmHg)
    Molecular Formula C8H6ClO2P
    MDL Number MFCD01630851
    UN Number UN3129
    Synonym Di(2-furyl)chlorophosphine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: WATER-REACTIVE LIQUID, CORROSIVE, N.O.S.
    Quantity 1g
    CAS 181257-35-2
    Chemical Name or Material Chlorodi(2-furyl)phosphine
    Assay ≥98%

    Description

    Applications
    Chlorodi(2-furyl)phosphine is a reactant in the synthesis 2, 2 ft.-bis (di-2-furylphosphino)-1, 1 ft.-binaphthalene. Chlorodi(2-furyl)phosphine is a reactant that is used in the synthesis of centrostereogenic 1,3-bidentate ferrocenyldiphosphane ligands, Ferrocene-based chiral diphosphine, and phosphine containing amino acids. In addition, Chlorodi(2-furyl)phosphine can be used as a reactant for α-Arylation of aldehydes, stereoselective sigmatropic rearrangement of acyclic allylic phosphinites and hydrophosphination of functionalized alkenes

    Solubility
    Reacts with water.

    Notes
    Air & Moisture Sensitive. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents, water, moisture, bases.

    GHS H Statement
    H261-H314-H318
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P231+P232-P260-P303+P361+P353-P305+P351+P338-P405-P501a
    Handle under inert gas.
    Protect from moisture.
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit