Alfa Aesar™ Chlorodi(o-tolyl)phosphine, 98%

Manufacturer: Alfa Aesar™ H2589003

Quantity 1g

 View more versions of this product

Catalog No. AAH2589003

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 36042-94-1
    Molecular Formula C14H14ClP
    Formula Weight 248.69
    MDL Number MFCD04038733
    UN Number UN1760
    Synonym Di(o-tolyl)chlorophosphine; Chlorobis(2-methylphenyl)phosphine
    Chemical Name or Material Chlorodi(o-tolyl)phosphine

    Specifications

    Formula Weight 248.69
    UN Number UN1760
    Synonym Di(o-tolyl)chlorophosphine; Chlorobis(2-methylphenyl)phosphine
    Boiling Point 174° to 178°C (3mmHg)
    Quantity 1g
    Molecular Formula C14H14ClP
    MDL Number MFCD04038733
    CAS 36042-94-1
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, N.O.S.
    Melting Point 57°C
    Chemical Name or Material Chlorodi(o-tolyl)phosphine
    Assay 98%

    Description

    Applications
    Chlorodi(o-tolyl)phosphine is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dye stuff.

    Solubility
    Not miscible or difficult to mix in water.

    Notes
    Air and moisture sensitive. Incompatible with oxidizing agents, bases, water/ moisture.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit