Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorodifluoroacetic anhydride, 98+%

Manufacturer:  Alfa Aesar™ A1243230

Quantity 250g

 View more versions of this product

Catalog No. AAA1243230


  Check Availability
Add to cart

Specifications

Description

Applications
It is used as the reagent for the derivatization of amines, alcohols, etc., for GC purposes. It has been utilized for dual element acetylation tagging derivatization of amines by using microwave sustained helium emission detector1 and as a derivatization reagent in determination of estrone and 17β-estradiol in hair by GC-MS2.

Solubility
Slightly Soluble in water. (1.4 g/L) (25°C)

Notes
Moisture sensitive. Handle under dry protective gas. Keep away from oxidizing agents, strong bases.
Description & Specifications

Chemical Identifiers

2834-23-3
242.94
UN3265
Chlorodifluoroacetic anhydride
(ClCF2CO)2O
MFCD00000774
1912832

Specifications

92° to 93°C
2834-23-3
242.94
UN3265
Chlorodifluoroacetic anhydride
250g
(ClCF2CO)2O
MFCD00000774
1912832
≥98%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

TSCA : TSCA

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit