Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium, 97%, ACROS Organics™

Click to view available options
Quantity:
250mg
1g
Packaging:
Glass bottle
791629-96-4
C16H37NP2·ClH2Ir
535.1
QPAONQONOKDMMJ-UHFFFAOYSA-M
iridium 3+ bis 2-diisopropylphosphanyl ethyl amine chloride dihydride,iridium 3+ chloride hydride-2-di propan-2-yl phosphanyl-n-2-di propan-2-yl phosphanyl ethyl ethan-1-amine 1/1/2/1
791629-96-4
C16H37NP2·ClH2Ir
535.1
QPAONQONOKDMMJ-UHFFFAOYSA-M
iridium 3+ bis 2-diisopropylphosphanyl ethyl amine chloride dihydride,iridium 3+ chloride hydride-2-di propan-2-yl phosphanyl-n-2-di propan-2-yl phosphanyl ethyl ethan-1-amine 1/1/2/1
Reduction reagent
Quantity 250mg
Infrared Spectrum Authentic
Melting Point 240.6°C to 241.9°C
Color Tan to White
Packaging Glass bottle
Elemental Analysis 34.50 to 37.30% C, 2.50 to 2.70% N
Assay Percent Range 97%
Formula Weight 535.1
Physical Form Powder
Percent Purity 97%
Chemical Name or Material chlorodihydrido[bis[(2-diisopropylphosphino)ethyl]amine]iridium
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning