Alfa Aesar™ Chlorodimethyl-n-octadecylsilane, tech. 90%, cont. 5-10% branched isomers

Manufacturer: Alfa Aesar™ B2357814

Quantity 25g

 View more versions of this product

Catalog No. AAB2357814

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 18643-08-8
    Molecular Formula C20H43ClSi
    Formula Weight 347.1
    MDL Number MFCD00000504
    UN Number UN3261
    Beilstein 2246012
    Synonym Dimethyl-n-octadecylsilyl chloride; n-Octadecyldimethylchlorosilane
    Chemical Name or Material Chlorodimethyl-n-octadecylsilane
    Grade Technical Grade

    Specifications

    Formula Weight 347.1
    Quantity 25g
    TSCA TSCA
    Odor Acrid
    Molecular Formula C20H43ClSi
    MDL Number MFCD00000504
    CAS 18643-08-8
    UN Number UN3261
    Beilstein 2246012
    Synonym Dimethyl-n-octadecylsilyl chloride; n-Octadecyldimethylchlorosilane
    Boiling Point 184° to 186°C (0.2mmHg)
    Melting Point 28° to 32°C
    Flash Point 200°C (392°F)
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 1 Danger
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    Chemical Name or Material Chlorodimethyl-n-octadecylsilane
    Assay 90%, cont. 5 to 10% branched isomers
    Grade Technical Grade

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit