Learn More
Alfa Aesar™ Chlorodimethyl-n-octadecylsilane, tech. 90%, cont. 5-10% branched isomers
Specifications
UN Number | UN3261 |
Beilstein | 2246012 |
Formula Weight | 347.1 |
Percent Purity | 90% |
Grade | Technical |
Odor | Acrid |
Boiling Point | 184°C to 186°C (0.2mmHg) |
Melting Point | 28°C to 32°C |
Quantity | 5g |
Flash Point | 200°C (392°F) |
Show More |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Danger
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
TSCA : TSCA