Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorodimethylphenylsilane, 95%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
768-33-2
C8H11ClSi
170.72
MFCD00000499
KWYZNESIGBQHJK-UHFFFAOYSA-N
chlorodimethylphenylsilane,chlorodimethyl phenyl silane,phenyldimethylchlorosilane,silane, chlorodimethylphenyl,dimethylphenylchlorosilane,chloro dimethyl phenylsilane,phenyl dimethylchlorosilane,chloro-dimethyl-phenyl-silane,benzene, chlorodimethylsilyl,pubchem10814
13029
chloro-dimethyl-phenylsilane
C[Si](C)(C1=CC=CC=C1)Cl
768-33-2
C8H11ClSi
170.72
MFCD00000499
KWYZNESIGBQHJK-UHFFFAOYSA-N
chlorodimethylphenylsilane,chlorodimethyl phenyl silane,phenyldimethylchlorosilane,silane, chlorodimethylphenyl,dimethylphenylchlorosilane,chloro dimethyl phenylsilane,phenyl dimethylchlorosilane,chloro-dimethyl-phenyl-silane,benzene, chlorodimethylsilyl,pubchem10814
13029
chloro-dimethyl-phenylsilane
C[Si](C)(C1=CC=CC=C1)Cl
Boiling Point 80°C to 84°C (16.0mmHg)
Density 1.0320g/mL
Flash Point 61°C
Specific Gravity 1.032
Refractive Index 1.5060 to 1.51
Quantity 25g
Packaging Glass bottle
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 95%
Linear Formula C6H5Si(CH3)2Cl
Fieser 02,278
Solubility Information Solubility in water: may decompose
Formula Weight 170.72
Physical Form Liquid
Percent Purity ≥94%
Chemical Name or Material Chlorodimethylphenylsilane
Show More Show Less

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger