Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorodiphenylphosphine, 98+%

Click to view available options
Quantity:
10g
50g
250g
1079-66-9
C12H10ClP
220.636
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine,diphenylchlorophosphine,diphenylphosphinous chloride,phosphinous chloride, diphenyl,p-chlorodiphenylphosphine,diphenylphosphine chloride,phosphine, chlorodiphenyl,unii-wo975pjk1y,pph2cl,chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
1079-66-9
C12H10ClP
220.636
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine,diphenylchlorophosphine,diphenylphosphinous chloride,phosphinous chloride, diphenyl,p-chlorodiphenylphosphine,diphenylphosphine chloride,phosphine, chlorodiphenyl,unii-wo975pjk1y,pph2cl,chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
Flash Point 138°C (280°F)
Sensitivity Air and moisture sensitive
Refractive Index 1.636
Density 1.204 g/mL
Odor Acrid
Boiling Point 318°C to 321°C
Quantity 250g
Color Colorless to Yellow
Melting Point 14°C to 16°C
Linear Formula (C6H5)2PCl
UN Number UN3265
Beilstein 512032
Solubility Information Miscible with alcohol. Slightly miscible with ammonia.
Formula Weight 220.64
Physical Form Liquid
Percent Purity ≥98%
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P234-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P390-P501c

H290-H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

EINECSNumber : 214-093-2

TSCA : Yes

Recommended Storage : Ambient temperatures