Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

chlorodiphenylphosphine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
500g
Packaging:
Glass bottle
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine,diphenylchlorophosphine,diphenylphosphinous chloride,phosphinous chloride, diphenyl,p-chlorodiphenylphosphine,diphenylphosphine chloride,phosphine, chlorodiphenyl,unii-wo975pjk1y,pph2cl,chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
1079-66-9
C12H10ClP
220.64
MFCD00000529
XGRJZXREYAXTGV-UHFFFAOYSA-N
chlorodiphenylphosphine,diphenylchlorophosphine,diphenylphosphinous chloride,phosphinous chloride, diphenyl,p-chlorodiphenylphosphine,diphenylphosphine chloride,phosphine, chlorodiphenyl,unii-wo975pjk1y,pph2cl,chloro diphenyl phosphane
66180
chloro(diphenyl)phosphane
C1=CC=C(C=C1)P(C2=CC=CC=C2)Cl
Quantity 500g
Refractive Index 1.6340 to 1.6370
Packaging Glass bottle
Density 1.2290g/mL
Flash Point 138°C
Boiling Point 320.0°C
Color Colorless to Yellow
Specific Gravity 1.229
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Linear Formula (C6H5)2PCl
Beilstein 16, 763
Fieser 11,120; 16,77
Merck Index 15, 2139
Solubility Information Solubility in water: reacts violently with water
Formula Weight 220.64
Physical Form Liquid
Percent Purity 98%
Chemical Name or Material Chlorodiphenylphosphine
Show More Show Less

GHS H Statement
May be corrosive to metals.
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Reacts violently with water.

GHS P Statement
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if pre

GHS Signal Word: Danger

EINECSNumber : 214-093-2

Recommended Storage : may become turbid on storage