SHOWA™ Chloroflex II™ Gloves

Neoprene gloves provide superior protection from petroleum-based materials

Manufacturer: SHOWA™ N811

 View more versions of this product

Catalog No. 19-120-2089C

  Check Availability
Add to Cart

    Specifications

    Certifications 21 CFR 170-199
    Glove Cuff Style Rolled
    For Use With (Application) Oil spill clean up, gear assembly, transformer intenance, agricultural chemicals
    Length (English) 18 in.
    Length (Metric) 45.72cm
    Lining Unlined
    Material Neoprene
    Size 11
    Thickness 30mil
    Coating Material Bisque
    Color Black

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit