Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, 99.8%, for analysis, stabilized with amylene, ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
25L
67-66-3,513-35-9
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3,513-35-9
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
For analysis
Acidity Alkalinity 0.0005 meq/g max.
Assay Percent Range 99.8%
Linear Formula CHCl3
Merck Index 15, 2142
Solubility Information Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Calcium (Ca) 0.1ppm max.
Formula Weight 119.38
Physical Form Liquid
Percent Purity 99.8%
Carbon Tetrachloride < 0.005%
Grade Analysis
Copper (Cu) 0.02ppm max.
Density 1.4800g/mL
Dichloromethane < 0.005%
Free Chlorine 5ppm max.
Iron (Fe) 0.1ppm max.
Lead (Pb) 0.02ppm max.
Magnesium (Mg) 0.05ppm max.
Potassium (K) 0.1ppm max.
Residue after Evaporation 5ppm max.
Silicon (Si) 0.05ppm max.
Solubility Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Sodium (Na) 0.2ppm max.
Specific Gravity 1.48
Stabilizer approx. 100ppm amylene
Substances Darkened by H2SO4 5 APHA max.
Suitability for use with dithizone: Passes Test
Tetrachloroethylene Tetrachloroethylene< 0.005%
Total Phosphorus 0.1ppm max.
Trace Analysis 0.05ppm max.
Trichloroethylene trichloroethylene < 0.005%
Total Sulfur 0.5ppm max.
Water 0.02% max. (K.F.)
Viscosity 0.56 mPa.s (20°C)
Boiling Point 61.0°C
Color Undesignated
Melting Point -63.0°C
Quantity 25L
Chemical Name or Material Chloroform, Stabilized with amylene
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning