Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, 99.9%, for residue analysis, ECD tested for pest anal., residue <0., ACROS Organics™

Click to view available options
Quantity:
1L
2.5L
Packaging:
Glass bottle
67-66-3, 513-35-9
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3, 513-35-9
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
For residue analysis
Additional Information GC/ECD (as lindane): max 5ng/L
Assay Percent Range 99.9%
Linear Formula CHCl3
Fieser 01,130; 08,92; 15,84
Formula Weight 119.38
Physical Form Liquid
Percent Purity 99.9%
Grade Residue Analysis
Density 1.4800g/mL
Hydrochloric Acid 0.001% max.
Packaging Glass bottle
Residue after Evaporation 0.0005% max.
Solubility Solubility in water: 8g/L (20°C). Other solubilities: miscible with alcohol, benzene, ether, petr.ether,, carbon tetrachloride, carbon disulfide and oils., soluble in acetone.
Specific Gravity 1.48
Stabilizer 0.0035 to 0.005%W/W (amylene)
Water 0.01% max. (Coulometric)
Viscosity 0.56 mPa.s (20°C)
Boiling Point 61.0°C
Color Undesignated
Melting Point -63.0°C
Quantity 1L
Chemical Name or Material Chloroform, ECD Tested, For Pesticide Analysis, Residue <0.0005%
Show More Show Less

GHS P Statement Causes skin irritation. Causes serious eye irritation. Harmful if swallowed. Toxic if inhaled. Suspected of causing cancer. May cause drowsiness or dizziness. Suspected of damaging the unborn child. Causes damage to organs through prolonged or repeated exposure if inhaled.

GHS P Statement IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician. IF ON SKIN: Wash with plenty of soap and water. Do not breathe dust/fume/gas/mist/vapors/spray. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 200-663-8

RTECSNumber : FS9100000

TSCA : TSCA