Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, ACS, >99.8%, Alfa Aesar™

Click to view available options
Quantity:
1L
4L
4 x 4L
67-66-3
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3
CHCl3
119.369
MFCD00000826
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
Assay Percent Range >99.8%
Beilstein 1731042
Solubility Information Miscible with diethyl ether,oils,ligroin,alcohol,carbon tetrachloride and carbon disulfide.
Formula Weight 119.38
Physical Form Liquid
Grade ACS Reagent
Density 1.492 g/mol
Refractive Index 1.446
Boiling Point 60.5°C to 61.5°C
Melting Point -63°C
Quantity 1L
Chemical Name or Material Chloroform
Show More Show Less

P201-P202-P260-P264b-P270-P271-P280i-P281-P301+P312-P302+P352-P304+P340-P305+P351+P338-P308+P313-P330-P332+P313-P362-P501c

H302-H315-H319-H331-H335-H336-H351-H361d-H373

DOTInformation : Hazard Class: 6.1; Packaging Group: III

EINECSNumber : 200-663-8

RTECSNumber : FS9100000

TSCA : Yes

Recommended Storage : Ambient temperatures