Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene, 1% ethanol, >99.9% (excluding ethanol), Honeywell Burdick & Jackson

Click to view available options
Quantity:
4 x 4L
6 x 1L
Packaging:
Glass Bottle
67-66-3, 64-17-5, 513-35-9
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3, 64-17-5, 513-35-9
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
Absorbance 0.3 AU max. at 250 nm (UV)
CAS Max % 98.9900%, 1.0000%, <0.0100%
UN Number UN1888
Beilstein 1731042
Solubility Information 81.5% in Water at 20°C
Formula Weight 119.37
Percent Purity >99.9%
Grade HPLC
Density 1.49 g/cm3
Packaging Glass Bottle
Solubility 81.5% in Water at 20°C
Water 0.02% max.
Boiling Point 62°C
Quantity 4 x 4L
Chemical Name or Material Chloroform, B&J Brand™, for HPLC, GC, pesticide residue analysis, and spectrophotometry, contains amylene and 1% ethanol, >99.9% (excluding ethanol)
Show More Show Less

ShelfLife : 547 days from date of manufacture

DOTInformation : Transport Hazard Class: 6.1; Packing Group:3: III; Proper Shipping Name: Chloroform