Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl

High quality chemicals for laboratory and specialized industrial use

    For Histological Use
Formula Weight 119.38
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth PVC Coated Amber Bottle
Specific Gravity 1L = 1.48kg
Quantity 4L
Chemical Name or Material Chloroform