Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
1L
4L
Packaging:
Aluminum Bottle
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Bottle

Chemical Identifiers

67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
CHEBI:35255

High quality chemicals for laboratory and specialized industrial use

    For Histological Use; For Laboratory, Research, or Manufacturing Use
SMILES C(Cl)(Cl)Cl
IUPAC Name chloroform
Formula Weight 119.38
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.48kg
Quantity 4L
Chemical Name or Material Chloroform
Grade Baker Analyzed™ ACS Reagent, Meets ACS reagent chemical requirements