Learn More
Chloroform-d, for NMR, 99.8+% atom D, contains 0.03 v/v% TMS, ACROS Organics™
Description
Specifications
Color | White to Beige |
Deuterium (D) | 99.80% min. |
Water | 0.01% max. (Coulometric) |
Packaging | Glass bottle |
Quantity | 100mL |
Density | 1.5000g/mL |
Boiling Point | 60.0°C |
Melting Point | -64.0°C |
Linear Formula | CDCl3 |
Beilstein | 01, IV, 54 |
Show More |
Safety and Handling
GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.
GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.
Warning
EINECSNumber : 212-742-4