Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform-d, for NMR, 99.8% atom D, stabilized with silver foil, ACROS Organics™

Click to view available options
Quantity:
25mL
100mL
500mL
865-49-6
CHCl3
120.375
MFCD00000827
HEDRZPFGACZZDS-MICDWDOJSA-N
chloroform-d,deuterochloroform,methane-d, trichloro,2h chloroform,deuterated chloroform,cdcl3,trichloromethane-d,unii-p1nw4885vt,trichloro deuterio methane,chloroform, deutero
71583
CHEBI:85365
trichloro(deuterio)methane
C(Cl)(Cl)Cl
865-49-6
CHCl3
120.375
MFCD00000827
HEDRZPFGACZZDS-MICDWDOJSA-N
chloroform-d,deuterochloroform,methane-d, trichloro,2h chloroform,deuterated chloroform,cdcl3,trichloromethane-d,unii-p1nw4885vt,trichloro deuterio methane,chloroform, deutero
71583
CHEBI:85365
trichloro(deuterio)methane
C(Cl)(Cl)Cl
Color Colorless
Hydrochloric Acid 0.001% max.
Deuterium (D) 99.75% min.
Chlorine (Cl) 0.0001% max.
Packaging Glass bottle
Water 0.02% max. (Coulometric)
Quantity 100mL
Density 1.5000g/mL
Boiling Point 60.0°C
Melting Point -64.0°C
Linear Formula CDCl3
Beilstein 01, IV, 54
Formula Weight 120.39
Physical Form Liquid
Grade NMR
Chemical Name or Material Chloroform-d, 99.8 atom% D
Show More Show Less

GHS H Statement
Harmful if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness. Suspected of damaging the unborn child.

GHS P Statement
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing. Call a POISON CENTER or doctor/physician if you feel unwell. IF ON SKIN: Wash with plenty of soap and water. Take off contaminated clothing and wash before reuse. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Wear protective gloves/protective clothing/eye protection/face protection.

Warning

EINECSNumber : 212-742-4