Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Histological Grade, 99.8%, Spectrum™

Click to view available options
Quantity:
1 L
4 L
20 L
Packaging:
Amber Glass Bottle
Steel Pail
67-66-3
67-66-3
CAS Max % 1
Percent Purity 0.998
Grade Histological
Free Chlorine To Pass Test
Packaging Amber Glass Bottle
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.