Chloroform, HPLC Grade, >99.5% min, Alfa Aesar™

Manufacturer: Alfa Aesar 22920K7

Quantity 4L

 View more versions of this product

Catalog No. AA22920K7

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 67-66-3
    Physical Form Liquid
    Assay Percent Range >99.5%
    Molecular Formula CHCl3
    Formula Weight 119.38
    MDL Number MFCD00000826
    Chemical Name or Material Chloroform
    Grade HPLC Grade

    Specifications

    Formula Weight 119.38
    Quantity 4L
    Color Colorless
    DOT Information DOT Class: 6.1; Packing Group: III
    EINECS Number 200-663-8
    Physical Form Liquid
    CAS 67-66-3
    Odor Characteristic
    Vapor Pressure 210 hPa (158mm Hg) at 20°C
    Molecular Formula CHCl3
    MDL Number MFCD00000826
    Health Hazard 1 H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.
    Appearance Colorless liquid
    Assay Percent Range >99.5%
    Chemical Name or Material Chloroform
    Grade HPLC Grade

    H351-H373-H302-H315: Suspected of causing cancer. May cause damage to organs through prolonged or repeated exposure. Harmful if swallowed. Causes skin irritation.

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit