Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChloroForm, HPLC, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle

Chemical Identifiers

67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
CHEBI:35255

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry; For Laboratory, Research, or Manufacturing Use
CAS 67-66-3
Molecular Formula CHCl3
InChI Key HEDRZPFGACZZDS-UHFFFAOYSA-N
SMILES C(Cl)(Cl)Cl
IUPAC Name chloroform
Molecular Weight (g/mol) 119.369
PubChem CID 6212
ChEBI CHEBI:35255
Formula Weight 119.38
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L1=.48kg
Quantity 1L
Chemical Name or Material Chloroform
Grade HPLC Grade, Meets ACS reagent chemical requirements
Show More Show Less