Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ChloroForm, HPLC, J.T.Baker™

Click to view available options
Quantity:
1L
4L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC Coated Amber Glass Bottle
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane,formyl trichloride,methane, trichloro,trichloroform,methane trichloride,methenyl trichloride,methyl trichloride,trichlormethan,chloroforme,cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl

High purity solvents for use in analytical and preparative separations

    For use in Liquid Chromatography (HPLC & UHPLC) & Spectrophotometry; For Laboratory, Research, or Manufacturing Use
Formula Weight 119.38
Grade Meets ACS reagent chemical requirements
Packaging Narrow Mouth PVC Coated Amber Glass Bottle
Specific Gravity 1L1=.48kg
Quantity 4L
Chemical Name or Material Chloroform