Chloroform: Isoamyl Alcohol, 24:1 OmniPur™ EMD Millipore

Manufacturer: EMD Millipore 3160450ML

Packaging Glass Bottle
Quantity 450mL

 View more versions of this product

Catalog No. 31-604-50ML

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    UN Number 1888
    Chemical Name or Material Chloroform: Isoamyl Alcohol

    Specifications

    Quantity 450mL
    Storage Information Store in accordance with local regulations. Store in a segregated and approved area. Store in original container, protected from direct sunlight. Eliminate all ignition sources. Separate from oxidizing materials. Keep container tightly closed and sealed until ready for use. Containers that have been opened must be carefully resealed and kept upright to prevent leakage.
    Packaging Glass Bottle
    Color Colorless
    UN Number 1888
    Chemical Name or Material Chloroform: Isoamyl Alcohol
    Assay 99.0% min (chloroform), 98.0% min (isoamyl alcohol)

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit