Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, J.T.Baker™

Click to view available options
Quantity:
1L
4L
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
trichloromethane, formyl trichloride, methane, trichloro, trichloroform, methane trichloride, methenyl trichloride, methyl trichloride, trichlormethan, chloroforme, cloroformio
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl
Formula Weight 119.38
Physical Form Liquid
Density 1.483g/cm3
Color Colorless
Quantity 1L
Chemical Name or Material Chloroform