Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Meets or Exceeds A.C.S. Specifications, SpectroSolv (TM), 99.9%, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
Packaging:
Amber Glass Bottle
67-66-3
67-66-3
CAS Max % 1
Acid and Chloride To Pass Test
Percent Purity 0.999
Grade High Purity Solvent
Free Chlorine To Pass Test
Lead (Pb) 0.05 ppm
Packaging Amber Glass Bottle
Residue after Evaporation 10 ppm
Stabilizer 0.5 - 1.0%
Quantity 500 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.