Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, PHOTREX™ Reagent, J.T.Baker™

Click to view available options
Quantity:
500mL
4L
Packaging:
Narrow Mouth Amber Glass Bottle

Chemical Identifiers

67-66-3
CHCl3
119.369
HEDRZPFGACZZDS-UHFFFAOYSA-N
6212
CHEBI:35255
chloroform
C(Cl)(Cl)Cl

High purity solvents characterized for use in UV, visible and IR spectrophotometry

    For Spectrophotometry
Formula Weight 119.38
Packaging Narrow Mouth Amber Glass Bottle
Specific Gravity 1L = 1.48kg
Quantity 4L
Chemical Name or Material Chloroform
Grade PHOTREX™ Reagent