Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Purified, Spectrum™

Click to view available options
Quantity:
500 mL
4 L
20 L
55 gal
Packaging:
Amber Glass Bottle
Drum
Steel Can
67-66-3
67-66-3
CAS Max % 1
Grade Purified
Packaging Steel Can
Quantity 20 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.