Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Spectrum™

Click to view available options
Quantity:
100 mL
500 mL
1 L
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
67-66-3
67-66-3
CAS Max % 1
Aldehyde and Ketone To Pass Test
Grade Reagent
Free Chlorine To Pass Test
Packaging Amber Glass Bottle
Specific Gravity at 25 degC 1.476 - 1.480
Quantity 100 mL

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.