Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloroform, Stabilized, Spectrophotometric Grade, ACS, 99.8%, Spectrum™

Click to view available options
Quantity:
500 mL
1 L
4 L
200 L
Packaging:
Amber Glass Bottle
Drum
67-66-3
67-66-3
CAS Max % 1
Acid and Chloride To Pass Test
Percent Purity 0.998
Grade Spectrophotometric
Free Chlorine To Pass Test
Lead (Pb) 0.05 ppm
Packaging Amber Glass Bottle
Residue after Evaporation 1e-05
Quantity 4 L

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.