Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorohydroquinone, 90%, Tech., ACROS Organics™

Click to view available options
Quantity:
5g
100g
Packaging:
Glass Bottle
Plastic bottle
615-67-8
C6H5ClO2
144.56
MFCD00002341
AJPXTSMULZANCB-UHFFFAOYSA-N
chlorohydroquinone,2-chlorohydroquinone,monochlorohydroquinone,chloroquinol,1,4-benzenediol, 2-chloro,2-chloro-1,4-benzenediol,hydroquinone, chloro,2-chloro-1,4-dihydroxybenzene,2,5-dihydroxychlorobenzene,1,4-dihydroxy-2-chlorobenzene
301
CHEBI:27675
2-chlorobenzene-1,4-diol
C1=CC(=C(C=C1O)Cl)O
615-67-8
C6H5ClO2
144.56
MFCD00002341
AJPXTSMULZANCB-UHFFFAOYSA-N
chlorohydroquinone,2-chlorohydroquinone,monochlorohydroquinone,chloroquinol,1,4-benzenediol, 2-chloro,2-chloro-1,4-benzenediol,hydroquinone, chloro,2-chloro-1,4-dihydroxybenzene,2,5-dihydroxychlorobenzene,1,4-dihydroxy-2-chlorobenzene
301
CHEBI:27675
2-chlorobenzene-1,4-diol
C1=CC(=C(C=C1O)Cl)O
Melting Point 101°C to 102°C
Boiling Point 263°C
Infrared Spectrum Authentic
Color Brown to Gray
Quantity 100g
Packaging Plastic bottle
Assay Percent Range 90%
Beilstein 06, 849
Solubility Information Solubility in water: soluble in water. Other solubilities: soluble in alcohol,benzene,slightly soluble in ether
Formula Weight 144.56
Physical Form Fine Crystalline Powder
Percent Purity 90%
Grade Technical
Chemical Name or Material Chlorohydroquinone, tech., 90%
Show More Show Less

GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for

GHS Signal Word: Danger

EINECSNumber : 210-442-8

RTECSNumber : MX4800000

TSCA : TSCA