Alfa Aesar™ Chloromethyl thiocyanate, 97%

Manufacturer: Alfa Aesar™ A1210809

Quantity 10g

 View more versions of this product

Catalog No. AAA1210809

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 3268-79-9
    Molecular Formula ClCH2SCN
    Formula Weight 107.56
    MDL Number MFCD00001831
    UN Number UN2810
    Beilstein 1098487
    Chemical Name or Material Chloromethyl thiocyanate

    Specifications

    Formula Weight 107.56
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    F INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Health Hazard 2 GHS H Statement
    H300-H310-H330-H227-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    MDL Number MFCD00001831
    Health Hazard 1 Danger
    RTECS Number XK8800000
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
    Boiling Point 185°C
    Molecular Formula ClCH2SCN
    CAS 3268-79-9
    UN Number UN2810
    Flash Point 83°C (181°F)
    Odor Pungent
    Beilstein 1098487
    Quantity 10g
    Chemical Name or Material Chloromethyl thiocyanate
    Assay 97%

    GHS H Statement
    H300-H310-H330-H227-H315-H319-H335
    Fatal if swallowed.
    Fatal in contact with skin.
    Fatal if inhaled.
    Combustible liquid.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    F INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Specific treatment is urgent (see ... on this label).
    Rinse mouth.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit