Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloromethyltrimethylsilane, 98+%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
2344-80-1
C4H11ClSi
122.667
MFCD00000878
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
chloromethyl trimethylsilane,silane, chloromethyl trimethyl,trimethylsilyl methyl chloride,chloromethyl trimethyl silane,ch3 3sich2cl,chloromethyl trimethylsilane,chloromethyl trimethyl silane,tmcms,pubchem13668,trimethylchloromethylsilane
75361
chloromethyl(trimethyl)silane
C[Si](C)(C)CCl
2344-80-1
C4H11ClSi
122.667
MFCD00000878
OOCUOKHIVGWCTJ-UHFFFAOYSA-N
chloromethyl trimethylsilane,silane, chloromethyl trimethyl,trimethylsilyl methyl chloride,chloromethyl trimethyl silane,ch3 3sich2cl,chloromethyl trimethylsilane,chloromethyl trimethyl silane,tmcms,pubchem13668,trimethylchloromethylsilane
75361
chloromethyl(trimethyl)silane
C[Si](C)(C)CCl
Alkylation reagent
Quantity 5g
Refractive Index 1.4165 to 1.4185 (20°C, 589nm)
Color Colorless
Infrared Spectrum Authentic
Assay Percent Range 98+%
Linear Formula (CH3)3SiCH2Cl
Beilstein 04, III, 1844
Fieser 12,118; 13,76
Formula Weight 122.67
Physical Form Liquid
Percent Purity ≥98.0%
Chemical Name or Material Chloromethyltrimethylsilane, 98+%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Highly flammable liquid and vapor.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 219-058-5

TSCA : TSCA