Alfa Aesar™ Chloropentaamminecobalt(III) chloride

Manufacturer: Alfa Aesar™ 1376509

Quantity 10g

 View more versions of this product

Catalog No. AA1376509

  N/A
Add to Cart
  Our pricing system is currently unavailable. You are viewing list price.
You can save item(s) to your list now to purchase when our pricing system is available.

    Chemical Identifiers

    CAS 13859-51-3
    Linear Formula [CoCl(NH3)5]Cl2
    Formula Weight 250.44
    MDL Number MFCD00064728
    Synonym Pentaamminechlorocobalt(III) chloride
    Chemical Name or Material Chloropentaamminecobalt(III) chloride

    Specifications

    Formula Weight 250.44
    Solubility Information Slightly soluble in water, soluble in conc; H2SO4; Insoluble in alcohol
    Physical Form Powder
    CAS 13859-51-3
    Health Hazard 3 GHS P Statement
    P280h-P262
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.
    Linear Formula [CoCl(NH3)5]Cl2
    MDL Number MFCD00064728
    Density 1.819g/mL
    Quantity 10g
    TSCA TSCA
    Synonym Pentaamminechlorocobalt(III) chloride
    Health Hazard 1 Warning
    Odor Odorless
    Health Hazard 2 GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.
    Color Purple
    Chemical Name or Material Chloropentaamminecobalt(III) chloride

    GHS H Statement
    H351-H317
    Suspected of causing cancer.
    May cause an allergic skin reaction.

    GHS P Statement
    P280h-P262
    Wear protective gloves/clothing.
    Do not get in eyes, on skin, or on clothing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit