Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloropentafluorobenzene, 99%, ACROS Organics™

Click to view available options
Quantity:
1g
344-07-0
C6ClF5
202.508
MFCD00000534
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
chloropentafluorobenzene,pentafluorochlorobenzene,benzene, chloropentafluoro,pentafluorophenyl chloride,chloroperfluorobenzene,ccris 2394,pentafluorochloro benzene,1-chloro-2,3,4,5,6-pentafluoro-benzene,benzene, 1-chloro-2,3,4,5,6-pentafluoro,pentafluorophenylchloride
9579
1-chloro-2,3,4,5,6-pentafluorobenzene
C1(=C(C(=C(C(=C1F)F)Cl)F)F)F
344-07-0
C6ClF5
202.508
MFCD00000534
KGCDGLXSBHJAHZ-UHFFFAOYSA-N
chloropentafluorobenzene,pentafluorochlorobenzene,benzene, chloropentafluoro,pentafluorophenyl chloride,chloroperfluorobenzene,ccris 2394,pentafluorochloro benzene,1-chloro-2,3,4,5,6-pentafluoro-benzene,benzene, 1-chloro-2,3,4,5,6-pentafluoro,pentafluorophenylchloride
9579
1-chloro-2,3,4,5,6-pentafluorobenzene
C1(=C(C(=C(C(=C1F)F)Cl)F)F)F
Packaging Glass bottle
Infrared Spectrum Authentic
Quantity 1g
Color Colorless
Refractive Index 1.4220 to 1.4240 (20°C, 589nm)
Assay Percent Range 99%
Linear Formula ClC6F5
Formula Weight 202.51
Physical Form Liquid
Percent Purity ≥98.5%
Chemical Name or Material Chloropentafluorobenzene, 99%
Show More Show Less

GHS H Statement:
Harmful if inhaled.

GHS P Statement:
Do not breathe dust/fume/gas/mist/vapors/spray.
Wear protective gloves/eye protection/face protection.
IF INHALED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 206-450-6

RTECSNumber : CZ1225500

TSCA : TSCA