Alfa Aesar™ Chlorophenol Red sodium salt, 0.04% w/v aq. soln.

Manufacturer: Alfa Aesar™ 38699AP

Quantity 500mL

 View more versions of this product

Catalog No. AA38699AP

  Check Availability
Add to Cart

    Chemical Identifiers

    Formula Weight 445.26
    MDL Number MFCD00151199
    Chemical Name or Material Chlorophenol Red sodium salt

    Specifications

    Formula Weight 445.26
    Quantity 500mL
    TSCA TSCA
    Concentration 0.04% w/v aq. soln.
    Physical Form Liquid
    MDL Number MFCD00151199
    Density 0.990g/mL
    Chemical Name or Material Chlorophenol Red sodium salt

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit