Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Chlorophenol Red sodium salt, 0.04% w/v aq. soln.

Click to view available options
Quantity:
100mL
500mL
6 x 100mL
1217203-49-0
MFCD00151199
1217203-49-0
MFCD00151199
Concentration 0.04% w/v aq. soln.
Formula Weight 445.26
Physical Form Liquid
Density 0.990 g/mL
Quantity 100mL
Chemical Name or Material Chlorophenol Red sodium salt

TSCA : Yes

Recommended Storage : Ambient temperatures