Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chlorophyll Extract, Spectrum™

Click to view available options
Quantity:
5 kg
Packaging:
Poly Pail
Packaging Poly Pail
Quantity 5 kg
CAS 479-61-8
CAS Max % 1
Grade Reagent

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.