Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Chloropyramine hydrochloride

Click to view available options
Quantity:
1g
6170-42-9
C16H21Cl2N3
326.265
MFCD00079009
VEYWWAGBHABATA-UHFFFAOYSA-N
chloropyramine hydrochloride,chloropyramine hcl,alergosan,nilfan,allergan s hydrochloride,chlorpiramin hydrochloride,halopyramine hydrochloride,chlorpyramin hcl,chlorpyramine hydrochloride,unii-fwa92z14nn
80311
N'-[(4-chlorophenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
CN(C)CCN(CC1=CC=C(C=C1)Cl)C2=CC=CC=N2.Cl
6170-42-9
C16H21Cl2N3
326.265
MFCD00079009
VEYWWAGBHABATA-UHFFFAOYSA-N
chloropyramine hydrochloride,chloropyramine hcl,alergosan,nilfan,allergan s hydrochloride,chlorpiramin hydrochloride,halopyramine hydrochloride,chlorpyramin hcl,chlorpyramine hydrochloride,unii-fwa92z14nn
80311
N'-[(4-chlorophenyl)methyl]-N,N-dimethyl-N'-pyridin-2-ylethane-1,2-diamine;hydrochloride
CN(C)CCN(CC1=CC=C(C=C1)Cl)C2=CC=CC=N2.Cl

Competitive reversible H-1 receptor antagonist

Color White
Quantity 1g
Merck Index 14,2162
Formula Weight 326.26
Physical Form Crystalline Powder
Chemical Name or Material Chloropyramine hydrochloride

EINECSNumber : 228-216-2

TSCA : No

Recommended Storage : Ambient temperatures